4-Fluorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluorphenylhydrazin Hydrochlorid, 1-(4-Fluorphenyl)hydrazin Hydrochlorid, 4-Fluorphenylhydrazinhydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

F14203-10G F14203-50G
SIALF14203-10GEA 85.9 EUR
SIALF14203-10G SIALF14203-50G
4-Fluorphenylhydraziniumchlorid, Sigma-Aldrich®
4-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
Molecular Weight: 162.59 g/mol
MDL: MFCD00012942
CAS-Nummer: 823-85-8
EINECS: 212-521-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR