Fluoranthen, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
F807-100G
F807-500G
F807-5G
SIALF807-100GEA
89.2
EUR
SIALF807-100G
SIALF807-500G
SIALF807-5G
Fluoranthen, Sigma-Aldrich®
Fluoranthene
Formel:
C₁₆H₁₀ Molecular Weight: 202,26 g/mol |
MDL:
MFCD00001184 CAS-Nummer: 206-44-0 EINECS: 205-912-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...