Methylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Aminomethan Hydrochlorid, Methylamin Hydrochlorid
M0505-100G
M0505-500G
SIALM0505-100GEA
28.8
EUR
SIALM0505-100G
SIALM0505-500G
Methylammoniumchlorid, Sigma-Aldrich®
Methylammonium chloride
Formel:
CH₆ClN Molecular Weight: 67.52 g/mol |
MDL:
MFCD00012849 CAS-Nummer: 593-51-1 EINECS: 209-795-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...