Isoamylcinnamat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
trans-3-Phenylpropensäureisoamylester, trans-Zimtsäureisopentylester, Isopentylcinnamat, trans-Zimtsäureisoamylester, (E)-3-Phenylpropensäureisoamylester
M-Clarity™ Qualitätsstufe = MQ300
W206318-10KG-K
W206318-1KG-K
SIALW206318-10KG-KEA
1420
EUR
SIALW206318-10KG-K
SIALW206318-1KG-K
Isoamylcinnamat, Sigma-Aldrich®
Isoamylcinnamat
Formel:
C₁₄H₁₈O₂ Molecular Weight: 218.3 g/mol |
CAS-Nummer:
7779-65-9 EINECS: 231-931-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...