Isoamylcinnamat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-3-Phenylpropensäureisoamylester, trans-Zimtsäureisopentylester, Isopentylcinnamat, trans-Zimtsäureisoamylester, (E)-3-Phenylpropensäureisoamylester

M-Clarity™ Qualitätsstufe = MQ300

W206326-100G-K W206326-1KG-K W206326-5KG-K
SIALW206326-100G-KEA 158 EUR
SIALW206326-100G-K SIALW206326-1KG-K SIALW206326-5KG-K
Isoamylcinnamat, Sigma-Aldrich®
Isoamylcinnamat
Formel: C₁₄H₁₈O₂
Molecular Weight: 218.3 g/mol
CAS-Nummer: 7779-65-9
EINECS: 231-931-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR