Isopropylcinnamat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
trans-3-Phenylpropensäureisopropylester, trans-Zimtsäureisopropylester, (E)-3-Phenylpropensäureisopropylester
M-Clarity™ Qualitätsstufe = MQ300
W293903-10KG-K
W293903-1KG-K
W293903-5KG-K
SIALW293903-10KG-KEA
1470
EUR
SIALW293903-10KG-K
SIALW293903-1KG-K
SIALW293903-5KG-K
Isopropylcinnamat, Sigma-Aldrich®
Isopropylcinnamat
Formel:
C₁₂H₁₄O₂ Molecular Weight: 190.24 g/mol |
MDL:
MFCD00048292 CAS-Nummer: 7780-06-5 EINECS: 231-949-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...