Allylchlordimethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Allyldimethylchlorsilan, ADMCS

SILS00225.5 SILS00225.25 SILS00225.100
SILNSILS00225.5EA 0 EUR
SILNSILS00225.5 SILNSILS00225.25 SILNSILS00225.100
Allylchlordimethylsilan
Allylchlordimethylsilan
Formel: C₅H₁₁ClSi
Molecular Weight: 134.68 g/mol
MDL: MFCD00000506
CAS-Nummer: 4028-23-3
EINECS: 223-706-2
UN: 2985
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR