(Chlormethyl)dimethylchlorsilan

Lieferant: CHEMPUR SILANE

Synonyme: CMDMCS, Chlor(chlormethyl)dimethylsilan

SILS04000.25 SILS04000.100
SILNSILS04000.25EA 0 EUR
SILNSILS04000.25 SILNSILS04000.100
(Chlormethyl)dimethylchlorsilan
(Chlormethyl)dimethylchlorsilan
Formel: C₃H₈Cl₂Si
Molecular Weight: 143,09 g/mol
MDL: MFCD00000875
CAS-Nummer: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR