Chlordimethylvinylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Dimethylvinylchlorsilan, Vinyldimethylchlorsilan

SILS21675.25 SILS21675.100
SILNSILS21675.25EA 0 EUR
SILNSILS21675.25 SILNSILS21675.100
Chlordimethylvinylsilan
Chlordimethylvinylsilan
Formel: C₄H₉ClSi
Molecular Weight: 120.65 g/mol
MDL: MFCD00018090
CAS-Nummer: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR