Dichlorethylvinylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Vinylethyldichlorsilan

SILS21750.10 SILS21750.25
SILNSILS21750.10EA 0 EUR
SILNSILS21750.10 SILNSILS21750.25
Dichlorethylvinylsilan
Dichlorethylvinylsilan
Formel: C₄H₈Cl₂Si
Molecular Weight: 155.1 g/mol
CAS-Nummer: 10138-21-3
EINECS: 233-384-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR