Dichlormethylvinylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Vinylmethyldichlorsilan

SILS21800.25 SILS21800.100 SILS21800.500
SILNSILS21800.25EA 0 EUR
SILNSILS21800.25 SILNSILS21800.100 SILNSILS21800.500
Dichlormethylvinylsilan
Dichlormethylvinylsilan
Formel: C₃H₆Cl₂Si
Molecular Weight: 141.07 g/mol
MDL: MFCD00000492
CAS-Nummer: 124-70-9
EINECS: 204-710-3
UN: 2985
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR