1-Brom-2-fluor-4-iodbenzol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A19190.06
A19190.14
A19190.22
A19190.06EA
57.6
EUR
A19190.06
A19190.14
A19190.22
1-Brom-2-fluor-4-iodbenzol ≥97%
1-Brom-2-fluor-4-iodbenzol
Purity: 97%
CAS No.: 136434-77-0
Molecular Formula: C6H3BrFI
Formula Weight: 300.90
Flash Point:
Formel:
C₆H₃BrFI Molecular Weight: 300,9 g/mol Siedepunkt: 80 °C (2,5 mmHg) Schmelzpunkt: 34…38 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042577 CAS-Nummer: 136434-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...