1-Brom-2-fluor-4-iodbenzol

Lieferant: FLUOROCHEM

008674-10G 008674-25G
FLUO008674-10GEA 0 EUR
FLUO008674-10G FLUO008674-25G
1-Brom-2-fluor-4-iodbenzol
1-Brom-2-fluor-4-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300.9 g/mol
MDL: MFCD00042577
CAS-Nummer: 136434-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR