1-Brom-3-fluor-4-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Brom-2-fluor-1-iodbenzol

A15884.14 A15884.22
A15884.14EA 56.2 EUR
A15884.14 A15884.22
1-Brom-3-fluor-4-iodbenzol ≥98%
1-Brom-3-fluor-4-iodbenzol

Purity: 98% CAS No.: 105931-73-5 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Melting Point: 46-50° Beilstein Reference: 4740250 MDL No.: MFCD00010608

Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
Siedepunkt: 241 °C (1013 hPa)
Schmelzpunkt: 46…50 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010608
CAS-Nummer: 105931-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR