1-Brom-3-fluor-4-iodbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Brom-2-fluor-1-iodbenzol
001802-25G
001802-100G
FLUO001802-25GEA
30.8
EUR
FLUO001802-25G
FLUO001802-100G
1-Brom-3-fluor-4-iodbenzol
1-Brom-3-fluor-4-iodbenzol
Formel:
C₆H₃BrFI Molecular Weight: 300.9 g/mol Siedepunkt: 241 Schmelzpunkt: 46…49 °C |
MDL:
MFCD00010608 CAS-Nummer: 105931-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...