1-Brom-3-(trifluormethoxy)benzol
Lieferant: Thermo Scientific
Synonyme:
3-(Trifluoromethoxy)bromobenzene
TL00635.1
TL00635.10
TL00635.50
MAYBTL00635.1EA
0
EUR
MAYBTL00635.1
MAYBTL00635.10
MAYBTL00635.50
1-Brom-3-(trifluormethoxy)benzol
1-Brom-3-(trifluormethoxy)benzol
Formel:
C₇H₄BrF₃O Molecular Weight: 241,01 g/mol Siedepunkt: 78…81 °C (55 mmHg) Dichte: 1,635 g/cm³ (20 °C) |
MDL:
MFCD00040943 CAS-Nummer: 2252-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...