2-Brom-4-fluorphenol ≥98%

Lieferant: Thermo Scientific
Achtung

A11263.06 A11263.14 A11263.22 AB114209.0100
A11263.06EA 37 EUR
A11263.06 A11263.14 A11263.22 ABCRAB114209.0100
2-Brom-4-fluorphenol ≥98%
2-Brom-4-fluorphenol

Purity: 98+% CAS No.: 496-69-5 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Flash Point: 85°(185°F) Melting Point: 42-46° Boiling Point: 123-125°/50mm Beilstein Reference: 2355593 MDL No.: MFCD00010614

Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
Siedepunkt: 123…125 °C (50 mmHg)
Schmelzpunkt: 42…46 °C
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010614
CAS-Nummer: 496-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR