2-Brom-4-fluorphenol

Lieferant: FLUOROCHEM
Achtung

004231-100G 004231-500G 004231-5G
FLUO004231-100GEA 148 EUR
FLUO004231-100G FLUO004231-500G FLUO004231-5G
2-Brom-4-fluorphenol
2-Brom-4-fluorphenol
Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
MDL: MFCD00010614
CAS-Nummer: 496-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR