1-Brom-2-iodbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
001124-100G
001124-25G
FLUO001124-100GEA
180
EUR
FLUO001124-100G
FLUO001124-25G
1-Brom-2-iodbenzol
1-Brom-2-iodbenzol
Formel:
C₆H₄BrI Molecular Weight: 282,91 g/mol Siedepunkt: 250 °C (1013 hPa) Dichte: 2,2571 g/cm³ (25 °C) |
MDL:
MFCD00001030 CAS-Nummer: 583-55-1 EINECS: 209-508-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...