1-Brom-4-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Bromo-1-iodobenzene

A13859.09 A13859.18 A13859.30
A13859.09EA 54.8 EUR
A13859.09 A13859.18 A13859.30
1-Brom-4-iodbenzol ≥98%
1-Brom-4-iodbenzol

Purity: 98% CAS No.: 589-87-7 Molecular Formula: C6H4BrI Formula Weight: 282.91 Odour: Recognizable Melting Point: 89-93° Boiling Point: 252° Beilstein Reference: 1904545 MDL No.: MFCD00001051

Formel: C₆H₄BrI
Molecular Weight: 282,91 g/mol
Siedepunkt: 252 °C
Schmelzpunkt: 89…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001051
CAS-Nummer: 589-87-7
EINECS: 209-662-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR