1-Brom-4-iodbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Bromo-1-iodobenzene

005440.50G 005440-100G 005440-25G
FLUO005440.50GEA 54.9 EUR
FLUO005440.50G FLUO005440-100G FLUO005440-25G
1-Brom-4-iodbenzol
1-Brom-4-iodbenzol
Formel: C₆H₄BrI
Molecular Weight: 282.91 g/mol
Siedepunkt: 245
Lagertemperatur: Raumtemperatur
MDL: MFCD00001051
CAS-Nummer: 589-87-7
EINECS: 209-662-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR