4-Bromanilin ≥98%

Lieferant: Thermo Scientific
Gefahr

A15126.0B A15126.18 A15126.30
A15126.0BEA 388 EUR
A15126.0B A15126.18 A15126.30
4-Bromanilin ≥98%
4-Bromanilin

Purity: 98+% CAS No.: 106-40-1 Molecular Formula: C6H6BrN Formula Weight: 172.03 Physical Form: Cryst solid Odour: Characteristic Boiling Point: 222-224° Beilstein Reference: 742031 Merck Reference: 14,1404 UN No.: 2811 MDL No.: MFCD00007822

Formel: C₆H₆BrN
Molecular Weight: 172,02 g/mol
Siedepunkt: 222…224 °C
Schmelzpunkt: 60…65 °C
Dichte: 1,799 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007822
CAS-Nummer: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,1404

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR