2-Brom-5-nitroanisol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25331.04
B25331.09
B25331.04EA
39.8
EUR
B25331.04
B25331.09
2-Brom-5-nitroanisol ≥98%
2-Brom-5-nitroanisol
Purity: 98%
CAS No.: 77337-82-7
Molecular Formula: C7H6BrNO3
Formula Weight: 232.03
Melting Point: 102-107°
Beilstein Reference: 2520496
Formel:
C₇H₆BrNO₃ Molecular Weight: 232.03 g/mol Schmelzpunkt: 102…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041250 CAS-Nummer: 77337-82-7 EINECS: 278-669-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...