2,4-Dibromanisol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1,3-Dibrom-4-methoxybenzol
003063-100G
003063-500G
FLUO003063-100GEA
87.9
EUR
FLUO003063-100G
FLUO003063-500G
2,4-Dibromanisol
2,4-Dibromanisol
Formel:
C₇H₆Br₂O Molecular Weight: 265,93 g/mol Siedepunkt: 294 °C (1013 hPa) Schmelzpunkt: 60…61 °C |
MDL:
MFCD00000079 CAS-Nummer: 21702-84-1 EINECS: 244-536-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...