4,5-Dibromveratrol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1,2-Dibromo-4,5-dimethoxybenzene, 4,5-Dibromoveratrole

L01999.09 L01999.18 L01999.30
L01999.09EA 47.2 EUR
L01999.09 L01999.18 L01999.30
4,5-Dibromveratrol ≥98%
4,5-Dibromveratrol

Purity: 98+% CAS No.: 37895-73-1 Molecular Formula: C8H8Br2O2 Formula Weight: 295.96 Melting Point: 90-92° Beilstein Reference: 1953148

Formel: C₈H₈Br₂O₂
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014894
CAS-Nummer: 37895-73-1
EINECS: 253-705-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR