4-Bromveratrol ≥97%
Lieferant: Thermo Scientific
Synonyme:
1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene
A11133.14
A11133.22
AB108943.0100
A11133.14EA
57.4
EUR
A11133.14
A11133.22
ABCRAB108943.0100
4-Bromveratrol ≥97%
4-Bromveratrol
Purity: 97%
CAS No.: 2859-78-1
Molecular Formula: C8H9BrO2
Formula Weight: 217.06
Flash Point: 109°(228°F)
Refractive Index: 1.5730
Boiling Point: 255-256°
Beilstein Reference: 2046884
MDL No.: MFCD00008381
Formel:
C₈H₉BrO₂ Molecular Weight: 217,06 g/mol Siedepunkt: 255…256 °C Dichte: 1,510 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008381 CAS-Nummer: 2859-78-1 EINECS: 220-677-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...