4-Bromveratrol
Lieferant: CHEMPUR
Synonyme:
1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene
SB01150.250
SB01150.1
CHMPSB01150.250EA
0
EUR
CHMPSB01150.250
MAYBSB01150.1
4-Bromveratrol
4-Bromveratrol
Formel:
C₈H₉BrO₂ Molecular Weight: 217,06 g/mol Siedepunkt: 255 °C (1013 hPa) Dichte: 1,5068 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008381 CAS-Nummer: 2859-78-1 EINECS: 220-677-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...