2-Brom-1,3,5-trimethoxybenzol

Lieferant: FLUOROCHEM

008951-1G 008951-5G 008951-25G
FLUO008951-1GEA 60.9 EUR
FLUO008951-1G FLUO008951-5G FLUO008951-25G
2-Brom-1,3,5-trimethoxybenzol
2-Brom-1,3,5-trimethoxybenzol
Formel: C₉H₁₁BrO₃
MDL: MFCD00040757
CAS-Nummer: 1131-40-4
EINECS: 214-464-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR