5-Brom-1,2,3-trimethoxybenzol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Brom-3,4,5-trimethoxybenzol
B24872.06
B24872.14
B24872.22
B24872.06EA
38.7
EUR
B24872.06
B24872.14
B24872.22
5-Brom-1,2,3-trimethoxybenzol ≥97%
5-Brom-1,2,3-trimethoxybenzol
Purity: 97%
CAS No.: 2675-79-8
Molecular Formula: C9H11BrO3
Formula Weight: 247.10
Melting Point: 77-80°
MDL No.: MFCD00017169
Formel:
C₉H₁₁BrO₃ Molecular Weight: 247,09 g/mol Schmelzpunkt: 77…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017169 CAS-Nummer: 2675-79-8 EINECS: 220-223-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...