2,4,6-Tribromphenol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A12424.0B
A12424.30
A12424.0BEA
161
EUR
A12424.0B
A12424.30
2,4,6-Tribromphenol ≥98%
2,4,6-Tribromphenol
Purity: 98%
CAS No.: 118-79-6
Molecular Formula: C6H3Br3O
Formula Weight: 330.80
Melting Point: 86-92°
Boiling Point: 282-290° subl.
Merck Reference: 14,9613
UN No.: 3077
MDL No.: MFCD00002150
Formel:
C₆H₃Br₃O Molecular Weight: 330,8 g/mol Siedepunkt: 282…290 °C (Subl.) Schmelzpunkt: 86…92 °C Dichte: 2,550 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002150 CAS-Nummer: 118-79-6 EINECS: 204-278-6 ADR: 9,III Merck Index: 14,9613 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...