2-Bromthiophenol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Brom-2-mercaptobenzol, 2-Brombenzol-1-thiol, o-Bromthiophenol, 2-Brombenzolthiol, 2-Bromphenylmercaptan

B25537.06 B25537.14
B25537.06EA 59.1 EUR
B25537.06 B25537.14
2-Bromthiophenol ≥97%
2-Bromthiophenol

Purity: 97% CAS No.: 6320-02-1 Molecular Formula: C6H5BrS Formula Weight: 189.08 Flash Point: 110°(230°F) Refractive Index: 1.6350 Odour: Unpleasant Boiling Point: 114-116°/10mm Beilstein Reference: 2039769 UN No.: 2922 MDL No.: MFCD00004827

Formel: C₆H₅BrS
Molecular Weight: 189.08 g/mol
Siedepunkt: 114…116 °C (10 mmHg)
Dichte: 1.604
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004827
CAS-Nummer: 6320-02-1
EINECS: 228-665-4
UN: 2922
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR