4-(Diethylamino)benzaldehyd 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Diethylaminobenzaldehyde
A11825.22
A11825.36
A11825.22EA
42.5
EUR
A11825.22
A11825.36
4-(Diethylamino)benzaldehyd 97%
4-(Diethylamino)benzaldehyd
Purity: 97%
CAS No.: 120-21-8
Molecular Formula: C11H15NO
Formula Weight: 177.25
Flash Point: 165°(329°F)
Melting Point: 39-41°
Boiling Point: 173-175°/7mm
Beilstein Reference: 511102
MDL No.: MFCD00003382
Formel:
C₁₁H₁₅NO Molecular Weight: 177,25 g/mol Siedepunkt: 173…175 °C (7 mmHg) Schmelzpunkt: 39…41 °C Dichte: 1,03 g/cm³ (20 °C) Flammpunkt: 165 °C (329 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003382 CAS-Nummer: 120-21-8 EINECS: 204-377-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...