4-(Dimethylamino)benzaldehyd ACS

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(N,N-Dimethylamino)benzaldehyd, Ehrlich's Reagenz

36684.14 36684.22 36684.36
36684.14EA 33.4 EUR
36684.14 36684.22 36684.36
4-(Dimethylamino)benzaldehyd ACS
4-(Dimethylamino)benzaldehyd

CAS No.: 100-10-7 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 164°(327°F) Physical Form: Crystalline powder Sensitivity: Air and Light Sensitive Appearance: White to pale yellow Odour: Strong Melting Point: 73-75° Boiling Point: 176-177°/17mm Solubility: Slightly soluble in water. Soluble in alcohol, ether, chloroform, acetic acid Beilstein Reference: 606802 Merck Reference: 14,3230 MDL No.: MFCD00003381

Formel: (CH₃)₂NC₆H₄CHO
Molecular Weight: 149,19 g/mol
Siedepunkt: 176…177 °C (17 mmHg)
Schmelzpunkt: 73…75 °C
Dichte: 1,0254 g/cm³ (99.9 °C)
Flammpunkt: 164 °C (327 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003381
CAS-Nummer: 100-10-7
EINECS: 202-819-0
Merck Index: 14,3230
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR