3-Nitrosalicylaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxy-3-nitrobenzaldehyde

B23658.03 B23658.06
B23658.03EA 86.9 EUR
B23658.03 B23658.06
3-Nitrosalicylaldehyd ≥98%
3-Nitrosalicylaldehyd

Purity: 98% CAS No.: 5274-70-4 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 109-110° MDL No.: MFCD00041874

Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
Schmelzpunkt: 109…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041874
CAS-Nummer: 5274-70-4
EINECS: 226-098-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR