5-Nitrosalicylaldehyd 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxy-5-nitrobenzaldehyde

416180050 416180250
ACRO416180050EA 50.6 EUR
ACRO416180050 ACRO416180250
5-Nitrosalicylaldehyd 98+%
5-Nitrosalicylaldehyd
Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007337
CAS-Nummer: 97-51-8
EINECS: 202-587-0

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Powder or crystals
Infrared spectrum Conforms
Melting point 124 to 128 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR