3,5-Dinitrosalicylaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

B20424.03 B20424.06
B20424.03EA 53.8 EUR
B20424.03 B20424.06
3,5-Dinitrosalicylaldehyd ≥98%
3,5-Dinitrosalicylaldehyd

Purity: 98% CAS No.: 2460-59-5 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 71-74° Beilstein Reference: 1121900

Formel: C₇H₄N₂O₆
Molecular Weight: 212,12 g/mol
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007103
CAS-Nummer: 2460-59-5
EINECS: 219-551-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR