m-Anisaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Methoxybenzaldehyde

A13962.14 A13962.22
A13962.14EA 32.3 EUR
A13962.14 A13962.22
m-Anisaldehyd ≥98%
m-Anisaldehyd

Purity: 98% CAS No.: 591-31-1 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 121°(250°F) Refractive Index: 1.5530 Odour: Characteristic Boiling Point: 231-233° Beilstein Reference: 606013 MDL No.: MFCD00003361

Formel: C₈H₈O₂
Molecular Weight: 136.15 g/mol
Siedepunkt: 231…233 °C
Dichte: 1.118
Flammpunkt: 121 °C (250 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003361
CAS-Nummer: 591-31-1
EINECS: 209-712-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR