4-Ethoxybenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

A11987.14 A11987.22 A11987.36
A11987.14EA 28.2 EUR
A11987.14 A11987.22 A11987.36
4-Ethoxybenzaldehyd ≥97%
4-Ethoxybenzaldehyd

Purity: 97+% CAS No.: 10031-82-0 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 110°(230°F) Refractive Index: 1.5590 Melting Point: 13-14° Boiling Point: 253-255° Beilstein Reference: 386863 MDL No.: MFCD00003388

Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 253…255 °C
Schmelzpunkt: 13…14 °C
Dichte: 1,08
Flammpunkt: <110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003388
CAS-Nummer: 10031-82-0
EINECS: 233-093-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR