4-Octyloxybenzaldehyd ≥97%

Lieferant: Thermo Scientific

Synonyme: 4-(Octyloxy)benzaldehyde

B20692.06 B20692.14
B20692.06EA 46.4 EUR
B20692.06 B20692.14
4-Octyloxybenzaldehyd ≥97%
4-Octyloxybenzaldehyd

Purity: 97% CAS No.: 24083-13-4 Molecular Formula: C15H22O2 Formula Weight: 234.33 Refractive Index: 1.5220 Boiling Point: 140-142°/0.1mm MDL No.: MFCD00014136

Formel: C₁₅H₂₂O₂
Molecular Weight: 234,34 g/mol
Siedepunkt: 140…142 °C (0,1 mmHg)
Dichte: 0,982
Lagertemperatur: Raumtemperatur
MDL: MFCD00014136
CAS-Nummer: 24083-13-4
EINECS: 246-012-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR