6-Methoxysalicylaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Hydroxy-6-methoxybenzaldehyde

L13145.03 L13145.06
L13145.03EA 69.8 EUR
L13145.03 L13145.06
6-Methoxysalicylaldehyd ≥98%
6-Methoxysalicylaldehyd

Purity: 98+% CAS No.: 700-44-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 68-74° Beilstein Reference: 1101524

Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 68…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151830
CAS-Nummer: 700-44-7
EINECS: 211-844-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR