3-Hydroxy-p-anisaldehyd
Lieferant: FLUOROCHEM
Synonyme:
Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
011790-1KG
011790-25G
011790-100G
FLUO011790-1KGEA
0
EUR
FLUO011790-1KG
FLUO011790-25G
FLUO011790-100G
3-Hydroxy-p-anisaldehyd
3-Hydroxy-p-anisaldehyd
Formel:
C₈H₈O₃ Molecular Weight: 152.15 g/mol Siedepunkt: 308 Schmelzpunkt: 68…70 °C Dichte: 1.401 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003369 CAS-Nummer: 621-59-0 EINECS: 210-694-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...