2,3,4-Trimethoxybenzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10609.09 A10609.18 A10609.30
A10609.09EA 25.3 EUR
A10609.09 A10609.18 A10609.30
2,3,4-Trimethoxybenzaldehyd ≥98%
2,3,4-Trimethoxybenzaldehyde

Purity: 98+% CAS No.: 2103-57-3 Molecular Formula: C10H12O4 Formula Weight: 196.20 Flash Point: 110°(230°F) Refractive Index: 1.5547 Melting Point: 38-42° Boiling Point: 168-170°/12mm Beilstein Reference: 981091 MDL No.: MFCD00003310

Formel: C₁₀H₁₂O₄
Molecular Weight: 196.2 g/mol
Siedepunkt: 168…170 °C (12 mmHg)
Schmelzpunkt: 38…42 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003310
CAS-Nummer: 2103-57-3
EINECS: 218-271-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR