3,4-Dihydroxybenzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Protocatechualdehyd
A11558.06
A11558.14
A11558.22
A11558.06EA
45.9
EUR
A11558.06
A11558.14
A11558.22
3,4-Dihydroxybenzaldehyd ≥98%
3,4-Dihydroxybenzaldehyd
Purity: 98%
CAS No.: 139-85-5
Molecular Formula: C7H6O3
Formula Weight: 138.12
Melting Point: 152-157°
Beilstein Reference: 774381
Merck Reference: 14,7893
MDL No.: MFCD00003370
Formel:
C₇H₆O₃ Molecular Weight: 138,12 g/mol Siedepunkt: 295 °C (1013 hPa) Schmelzpunkt: 152…157 °C Dichte: 1,505 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003370 CAS-Nummer: 139-85-5 EINECS: 205-377-7 Merck Index: 14,7893 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...