2,3,4-Trihydroxybenzaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

A13745.06 A13745.14
A13745.06EA 49.9 EUR
A13745.06 A13745.14
2,3,4-Trihydroxybenzaldehyd ≥98%
2,3,4-Trihydroxybenzaldehyde

Purity: 98% CAS No.: 2144-08-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 159-163° Beilstein Reference: 2328658

Formel: C₇H₆O₄
Molecular Weight: 154,12 g/mol
Siedepunkt: 332 °C (1013 hPa)
Schmelzpunkt: 159…163 °C
Dichte: 1,054 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003325
CAS-Nummer: 2144-08-3
EINECS: 218-404-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR