2,4,5-Trihydroxybenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

A13283.03 A13283.06
A13283.03EA 125 EUR
A13283.03 A13283.06
2,4,5-Trihydroxybenzaldehyd ≥97%
2,4,5-Trihydroxybenzaldehyde

Purity: 97% CAS No.: 35094-87-2 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 230° dec. Beilstein Reference: 2250084

Formel: C₇H₆O₄
Molecular Weight: 154,12 g/mol
Schmelzpunkt: ∼230 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016592
CAS-Nummer: 35094-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR