2-Phenoxybenzaldehyd

Lieferant: BIONET RESEARCH
Achtung

11N-013.500 11N-013.1 11N-013.5 11N-013.10
BIOT11N-013.500EA 0 EUR
BIOT11N-013.500 BIOT11N-013.1 BIOT11N-013.5 BIOT11N-013.10
2-Phenoxybenzaldehyd
2-Phenoxybenzaldehyd
Formel: C₁₃H₁₀O₂
Molecular Weight: 198.22 g/mol
MDL: MFCD00800666
CAS-Nummer: 19434-34-5
EINECS: 243-061-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR