4-Nitrobenzonitril
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
4-Nitrobenzoesäurenitril
002987-25G
002987-100G
FLUO002987-25GEA
79.1
EUR
FLUO002987-25G
FLUO002987-100G
4-Nitrobenzonitril
4-Nitrobenzonitril
Formel:
C₇H₄N₂O₂ Molecular Weight: 148.12 g/mol Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007279 CAS-Nummer: 619-72-7 EINECS: 210-610-0 UN: 3439 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...