4-Methoxybenzonitril ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A15945.06 A15945.14 A15945.22
A15945.06EA 31 EUR
A15945.06 A15945.14 A15945.22
4-Methoxybenzonitril ≥99%
4-Methoxybenzonitril

Purity: 99% CAS No.: 874-90-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Odour: Almond-like Melting Point: 57-61° Boiling Point: 256-257° Beilstein Reference: 508117 UN No.: 3439 MDL No.: MFCD00001818

Formel: C₈H₇NO
Siedepunkt: 256…257 °C
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001818
CAS-Nummer: 874-90-8
EINECS: 212-871-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR