2,4-Dimethoxybenzonitril

Lieferant: CHEMPUR

TL00189.250 TL00189.1
CHMPTL00189.250EA 0 EUR
CHMPTL00189.250 MAYBTL00189.1
2,4-Dimethoxybenzonitril
2,4-Dimethoxybenzonitril
Formel: C₉H₉NO₂
Siedepunkt: 283 °C (1013 hPa)
MDL: MFCD00001786
CAS-Nummer: 4107-65-7
EINECS: 223-886-2
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR