3,5-Dihydroxybenzonitril ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L05642.03
L05642.06
L05642.03EA
67.8
EUR
L05642.03
L05642.06
3,5-Dihydroxybenzonitril ≥98%
3,5-Dihydroxybenzonitril
Purity: 98%
CAS No.: 19179-36-3
Molecular Formula: C7H5NO2
Formula Weight: 135.12
Melting Point: 185-190°
Beilstein Reference: 6322491
UN No.: 3439
MDL No.: MFCD00016453
Formel:
C₇H₅NO₂ Molecular Weight: 135,12 g/mol Siedepunkt: 311 °C (1013 hPa) Schmelzpunkt: 185…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016453 CAS-Nummer: 19179-36-3 EINECS: 242-859-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...